missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfanilamide 98+%, Reagents
Sulfanilamide 98+%, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2335200500A5
Specifications
| Sulfanilamide 98+% | |
| 165.5°C | |
| 165.5°C | |
| C6H8N2O2S | |
| 1.08 | |
| FDDDEECHVMSUSB-UHFFFAOYSA-N | |
| 4-aminobenzene-1-sulfonamide | |
| Reagent |
| 63-74-1 | |
| Colorless/White | |
| Poly Bottle | |
| 500 g | |
| Sulfamine, 4-Aminobenzenesulfonamide | |
| NC1=CC=C(C=C1)S(N)(=O)=O | |
| 172.20 | |
| Solid |
Chemical Identifiers
| 63-74-1 | |
| 172.20 | |
| Sulfamine, 4-Aminobenzenesulfonamide | |
| NC1=CC=C(C=C1)S(N)(=O)=O |
| C6H8N2O2S | |
| FDDDEECHVMSUSB-UHFFFAOYSA-N | |
| 4-aminobenzene-1-sulfonamide |