missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulfanilamide 98%

Sulfanilamide, CAS # 63-74-1, is an organic sulfur compound that shows antibacterial activities.

Supplier:  Thermo Scientific Chemicals 132855000

 View more versions of this product

Catalog No. AC132855000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sulfanilamide is an organic sulfur compound with antibacterial properties that are structurally similar to the p-aminobenzoic acid (PABA)
  • It also has a competitive inhibitory capacity against dihydropteroate synthetases blocking the synthesis of folic acid
  • Sulfanilamide competes with PABA for the bacterial enzyme dihydropteroate synthase preventing, the incorporation of PABA into dihydrofolic acid, an immediate precursor of folic acid. This culminates in the inhibition of the bacterial folic acid synthesis and also the de novo synthesis of purines and pyrimidines, resulting in cell growth arrest and cell death.

Applications

  • This compound shows antibacterial activity against different species, including gram-positive, gram-negative, and Chlamydia
  • In chemical reactions, this compound can act as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution
  • Sulfanilamide shows inhibitory activity against the enzyme, carbonic anhydrase.
Specifications

Specifications

Sulfanilamide
98%
63-74-1
Authentic
Plastic Bottle
500 g
C6H8N2O2S
MFCD00007939
15, 9057
Solubility in water: 7.5g/L (20°C).
C1=CC(=CC=C1N)S(=O)(=O)N
172.202
CHEBI:45373
98%
Sulfanilamide
Crystalline Powder
163.0°C to 167.0°C
0.5% max. (1 g, 105°C)
White
98%
4-(H2N)C6H4SO2NH2
14, 698
4-Aminobenzenesulfonamide
FDDDEECHVMSUSB-UHFFFAOYSA-N
4-aminobenzenesulfonamide
5333
172.2

Chemical Identifiers

63-74-1
172.202
FDDDEECHVMSUSB-UHFFFAOYSA-N
5333
4-aminobenzenesulfonamide
C6H8N2O2S
MFCD00007939
4-Aminobenzenesulfonamide
CHEBI:45373
C1=CC(=CC=C1N)S(=O)(=O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-563-4

RTECSNumber : WO8400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only