missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic acid, 99%
CAS: 5329-14-6 | H3NO3S | 97.09 g/mol
Supplier: Thermo Scientific Chemicals 222070025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sulfamic acid | |
| 200.0°C to 208.0°C | |
| Plastic Bottle | |
| 2.5 kg | |
| Crystals or Crystalline Powder | |
| 11117 | |
| amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid | |
| IIACRCGMVDHOTQ-UHFFFAOYSA-N | |
| sulfamic acid | |
| CHEBI:9330 |
| White | |
| 5329-14-6 | |
| 99% | |
| H3NO3S | |
| H2NSO3H | |
| 159053 | |
| Solubility in water: 146.8g/L (20°C). Other solubilities: sparingly soluble in alcohol and methanol,slightly soluble in acetone,insoluble in ether | |
| NS(=O)(=O)O | |
| 97.09 | |
| 99% |
Chemical Identifiers
| 5329-14-6 | |
| 97.09 | |
| amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid | |
| sulfamic acid |
| H3NO3S | |
| IIACRCGMVDHOTQ-UHFFFAOYSA-N | |
| CHEBI:9330 | |
| NS(=O)(=O)O |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 226-218-8
RUO – Research Use Only