missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic acid, 99%

Catalog No. p-4510161
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222072500 250 g Plastic Bottle
AC222070010 1 kg Plastic Bottle
AC222070025 2.5 kg Plastic Bottle
AC222070100 10 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC222072500 Supplier Thermo Scientific Chemicals Supplier No. 222072500
Only null left

CAS: 5329-14-6 | H3NO3S | 97.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.09
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

Color White
Melting Point 200.0°C to 208.0°C
Packaging Plastic Bottle
Assay Percent Range 99%
Quantity 250 g
Physical Form Crystals or Crystalline Powder
Linear Formula H2NSO3H
Fieser 11117
Merck Index 159053
Solubility Information Solubility in water: 146.8g/L (20°C). Other solubilities: sparingly soluble in alcohol and methanol,slightly soluble in acetone,insoluble in ether
Percent Purity 99%
Chemical Name or Material Sulfamic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 226-218-8

RUO – Research Use Only