missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, 10% w/v, Reagents

Catalog No. 01333519
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333519 20 L
01333520 10 L
01333518 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01333519 Supplier Labchem Inc Supplier No. CS21000020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfamic Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 5329-14-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.05
Quantity 20 L
Physical Form Liquid
Percent Purity 10% w/v
Grade Reagent
Chemical Name or Material Sulfamic Acid, 10% w/v