missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethazine 98.0+%, TCI America™
Supplier: TCI America S0586250G
Specifications
| Sulfamethazine | |
| 199°C | |
| C12H14N4O2S | |
| 250 g | |
| ASWVTGNCAZCNNR-UHFFFAOYSA-N | |
| 4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide | |
| 5327 | |
| 278.33 | |
| Crystalline Powder |
| 57-68-1 | |
| White | |
| MFCD00006066 | |
| sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin | |
| CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C | |
| 278.33 | |
| CHEBI:102265 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 57-68-1 | |
| 278.33 | |
| ASWVTGNCAZCNNR-UHFFFAOYSA-N | |
| 5327 | |
| 4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide |
| C12H14N4O2S | |
| MFCD00006066 | |
| sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin | |
| CHEBI:102265 | |
| CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C |
Safety and Handling
RTECSNumber : WO9275000
TSCA : Yes
Recommended Storage : Refrigerator