missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethazine 98.0+%, TCI America™

Catalog No. p-7135931
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S058625G 25 g
S0586250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S058625G Supplier TCI America Supplier No. S058625G
Only null left

Chemical Identifiers

CAS 57-68-1
Molecular Formula C12H14N4O2S
Molecular Weight (g/mol) 278.33
MDL Number MFCD00006066
InChI Key ASWVTGNCAZCNNR-UHFFFAOYSA-N
Synonym sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin
PubChem CID 5327
ChEBI CHEBI:102265
IUPAC Name 4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide
SMILES CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C

Specifications

Melting Point 199°C
Color White
Quantity 25 g
Formula Weight 278.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfamethazine
RTECSNumber WO9275000
TSCA Yes
Recommended Storage Refrigerator