missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethazine Sodium Salt 97.0+%, TCI America™

Catalog No. S058325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S058325G 25 g
S0583500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S058325G Supplier TCI America Supplier No. S058325G

Chemical Identifiers

CAS 1981-58-4
Molecular Formula C12H13N4NaO2S
Molecular Weight (g/mol) 300.312
MDL Number MFCD00068333
InChI Key NGIVTUVVBWOTNT-UHFFFAOYSA-N
Synonym sulfamethazine sodium salt, sulfamethazine sodium, sulfadimidine sodium, sodium sulfamethazine, sodium sulfametazine, sodium sulfamethamide, sodium sulfamezathine, bovibol, vesadin, sodium sulfamethiazine
PubChem CID 13456556
IUPAC Name sodium;(4-aminophenyl)sulfonyl-(4,6-dimethylpyrimidin-2-yl)azanide
SMILES CC1=CC(=NC(=N1)[N-]S(=O)(=O)C2=CC=C(C=C2)N)C.[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 300.31
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Sulfamethazine Sodium Salt
RTECSNumber WO9300000
TSCA Yes
Recommended Storage Refrigerator