accessibility menu, dialog, popup

UserName

NaCH3CH3NNSOOH2NN

CAS RN 1981-58-4

IUPAC Name: sodium 4-{[(4,6-dimethylpyrimidin-2-yl)azanidyl]sulfonyl}aniline
Synonyms: sodium 4-[(4,6-dimethylpyrimidin-2-yl)azanidylsulfonyl]aniline
Molecular Weight (g/mol): 300.31
Molecular Formula: C12H13N4NaO2S
InChi Key: NGIVTUVVBWOTNT-UHFFFAOYSA-N
SMILES: [Na+].CC1=CC(C)=NC([N-]S(=O)(=O)C2=CC=C(N)C=C2)=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)