missing translation for 'onlineSavingsMsg'
Learn More

Sulfadoxine, 98%

Catalog No. AAJ6626806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ6626806 5 g Glass bottle
AAJ6626814 25 g Plastic bottle
AAJ6626822 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6626806 Supplier Thermo Scientific Chemicals Supplier No. J6626806

CAS: 2447-57-6 | C12H14N4O4S | 310.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2447-57-6
Molecular Formula C12H14N4O4S
Molecular Weight (g/mol) 310.33
MDL Number MFCD00792890
InChI Key PJSFRIWCGOHTNF-UHFFFAOYSA-N
Synonym sulfadoxine, sulforthomidine, sulphadoxine, fanasil, sulphormethoxine, sulfadoxin, 4-amino-n-5,6-dimethoxypyrimidin-4-yl benzenesulfonamide, sulfadoxina, sulfadoxinum, fanzil
PubChem CID 17134
ChEBI CHEBI:9329
IUPAC Name 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide
SMILES COC1=NC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1OC

Specifications

Packaging Glass bottle
Quantity 5 g
Beilstein 625453
Solubility Information Soluble in ethanol & ammonium hydroxide (9:1) at 20mg/ml
Formula Weight 310.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sulfadoxine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-504-9
RTECSNumber DA9500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only