accessibility menu, dialog, popup

UserName

CAS RN 2447-57-6

IUPAC Name: 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide
Synonyms: sulfadoxine sulphormethoxine sulfadoxine product sulfadoxine product (product) sulfadoxine roche brand solfadossina sulformetoxine fanasil sulfadoxina sulforthomidine
Molecular Weight (g/mol): 310.33
Molecular Formula: C12H14N4O4S
InChi Key: PJSFRIWCGOHTNF-UHFFFAOYSA-N
SMILES: COC1=NC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1OC
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)