missing translation for 'onlineSavingsMsg'
Learn More

Sulfadoxin 98.0+%, TCI America™

Catalog No. p-7135985
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08995G 5 g
S089925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08995G Supplier TCI America Supplier No. S08995G

Chemical Identifiers

CAS 2447-57-6
Molecular Formula C12H14N4O4S
Molecular Weight (g/mol) 310.33
MDL Number MFCD00792890
InChI Key PJSFRIWCGOHTNF-UHFFFAOYSA-N
Synonym sulfadoxine, sulforthomidine, sulphadoxine, fanasil, sulphormethoxine, sulfadoxin, 4-amino-n-5,6-dimethoxypyrimidin-4-yl benzenesulfonamide, sulfadoxina, sulfadoxinum, fanzil
PubChem CID 17134
ChEBI CHEBI:9329
IUPAC Name 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide
SMILES COC1=NC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1OC

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 5 g
Formula Weight 310.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfadoxin
RTECSNumber DA9500000
TSCA No