missing translation for 'onlineSavingsMsg'
Learn More

Sulfadimethoxine, ≥98.5% (TLC), MilliporeSigma™ Supelco™

Chemical structure: sulfonamide

$147.55 - $822.50

Specifications

CAS 122-11-2
Physical Form Neat
Linear Formula C12H14N4O4 S
MDL Number MFCD00057345
Synonym 4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012798
View Documents
Merck Emd Millipore
S700710G
10 g N/A
Only null left
 
111012799
View Documents
Merck Emd Millipore
S7007100G
100 g N/A
Only null left
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

122-11-2
C12H14N4O4 S
4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide
310.33
Analytical Standard
Neat
MFCD00057345
Analytical Standard
≥98.5% (TLC)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'