missing translation for 'onlineSavingsMsg'
Learn More

Sulfadiazine 99.0+%, TCI America™

Supplier:  TCI America S057925G

Catalog No. S057925G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfadiazine
256°C
C10H10N4O2S
25 g
SEEPANYCNGTZFQ-UHFFFAOYSA-N
4-amino-N-pyrimidin-2-ylbenzenesulfonamide
5215
250.28
Crystalline Powder
68-35-9
White-Yellow
MFCD00006065
sulfadiazine, sulphadiazine, sulfapyrimidine, sulfadiazin, sulfazine, sulfadiazene, liquadiazine, sulfapyrimidin, adiazin, adiazine
C1=CN=C(N=C1)NS(=O)(=O)C2=CC=C(C=C2)N
250.276
CHEBI:9328
≥99.0% (HPLC,T)

Chemical Identifiers

68-35-9
250.276
SEEPANYCNGTZFQ-UHFFFAOYSA-N
5215
4-amino-N-pyrimidin-2-ylbenzenesulfonamide
C10H10N4O2S
MFCD00006065
sulfadiazine, sulphadiazine, sulfapyrimidine, sulfadiazin, sulfazine, sulfadiazene, liquadiazine, sulfapyrimidin, adiazin, adiazine
CHEBI:9328
C1=CN=C(N=C1)NS(=O)(=O)C2=CC=C(C=C2)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : WP1925000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions