accessibility menu, dialog, popup

UserName

CAS RN 68-35-9

IUPAC Name: 4-amino-N-(pyrimidin-2-yl)benzene-1-sulfonamide
Synonyms: sulfadiazine lantrisul sulfatryl pecta-diazine microsulfon sulphadiazine sulfaloid sulfanilamidopyrimidine solfadiazina quadetts
Molecular Weight (g/mol): 250.28
Molecular Formula: C10H10N4O2S
InChi Key: SEEPANYCNGTZFQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CC=N1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)