missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfabenzamide 98.0+%, TCI America™
Supplier: TCI America S058225G
Specifications
| Sulfabenzamide | |
| 182°C | |
| C13H12N2O3S | |
| 25 g | |
| PBCZLFBEBARBBI-UHFFFAOYSA-N | |
| N-(4-aminophenyl)sulfonylbenzamide | |
| 5319 | |
| ≥98.0% (HPLC,T) |
| 127-71-9 | |
| White-Yellow | |
| MFCD00044890 | |
| sulfabenzamide, sulfabenzide, sulfabenzid, sulfabenzoylamide, n-sulfanilylbenzamide, n-sulfamylbenzamide, n-benzoylsulfanilamide, sulfamylbenzamide, sulfabenzamid, sulfabenzamida | |
| C1=CC=C(C=C1)C(=O)NS(=O)(=O)C2=CC=C(C=C2)N | |
| 276.31 | |
| 276.31 | |
| Crystalline Powder |
Chemical Identifiers
| 127-71-9 | |
| 276.31 | |
| PBCZLFBEBARBBI-UHFFFAOYSA-N | |
| 5319 | |
| C1=CC=C(C=C1)C(=O)NS(=O)(=O)C2=CC=C(C=C2)N |
| C13H12N2O3S | |
| MFCD00044890 | |
| sulfabenzamide, sulfabenzide, sulfabenzid, sulfabenzoylamide, n-sulfanilylbenzamide, n-sulfamylbenzamide, n-benzoylsulfanilamide, sulfamylbenzamide, sulfabenzamid, sulfabenzamida | |
| N-(4-aminophenyl)sulfonylbenzamide |
Safety and Handling
RTECSNumber : CV5802500
TSCA : Yes
Recommended Storage : Refrigerator