accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 127-71-9

NH2SOOHNO

CAS RN 127-71-9

IUPAC Name: N-(4-aminobenzenesulfonyl)benzamide
Synonyms: sulfabenzamide 磺胺苯酰
Molecular Weight (g/mol): 276.31
Molecular Formula: C13H12N2O3S
InChi Key: PBCZLFBEBARBBI-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(=O)(=O)NC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.