Learn More
Thermo Scientific Chemicals Sudan III
CAS: 85-86-9 | C22H16N4O | 352.397 g/mol
$57.53 - $496.05
Chemical Identifiers
| CAS | 85-86-9 |
|---|---|
| Molecular Formula | C22H16N4O |
| Molecular Weight (g/mol) | 352.397 |
| MDL Number | MFCD00003905 |
| InChI Key | HTPQPMPFXUWUOT-UHFFFAOYSA-N |
| Synonym | C.I. 26100; Solvent Red 23 |
| PubChem CID | 6789251 |
| IUPAC Name | 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one |
| SMILES | C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1831814
|
Thermo Scientific Chemicals
A1831814 |
25 g |
Each for $57.53
|
|
|||||
|
AAA1831822
|
Thermo Scientific Chemicals
A1831822 |
100 g |
Each for $142.99
|
|
|||||
|
AAA1831836
|
Thermo Scientific Chemicals
A1831836 |
500 g |
Each for $496.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 85-86-9 | |
| 352.397 | |
| HTPQPMPFXUWUOT-UHFFFAOYSA-N | |
| 6789251 | |
| C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43 |
| C22H16N4O | |
| MFCD00003905 | |
| C.I. 26100; Solvent Red 23 | |
| 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one |
Specifications
| ∼200°C (decomposition) | |
| C22H16N4O | |
| 931185 | |
| C.I. 26100; Solvent Red 23 | |
| C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43 | |
| 352.397 | |
| 352.4 | |
| Sudan III |
| 85-86-9 | |
| MFCD00003905 | |
| 14,8884 | |
| HTPQPMPFXUWUOT-UHFFFAOYSA-N | |
| 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one | |
| 6789251 | |
| 25 g |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 201-638-4
RTECSNumber : QK4250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only