missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan III

CAS: 85-86-9 | C22H16N4O | 352.397 g/mol

Supplier:  Thermo Scientific Chemicals A1831814

 View more versions of this product

Catalog No. AAA1831814


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sudan III
85-86-9
MFCD00003905
14,8884
HTPQPMPFXUWUOT-UHFFFAOYSA-N
1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
6789251
25 g
∼200°C (decomposition)
C22H16N4O
931185
C.I. 26100; Solvent Red 23
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43
352.397
352.4

Chemical Identifiers

85-86-9
352.397
HTPQPMPFXUWUOT-UHFFFAOYSA-N
6789251
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43
C22H16N4O
MFCD00003905
C.I. 26100; Solvent Red 23
1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 201-638-4

RTECSNumber : QK4250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only