missing translation for 'onlineSavingsMsg'
Learn More

Suberic acid, 99%

CAS: 505-48-6 | C8H14O4 | 174.20 g/mol

$105.49 - $1,009.75

Chemical Identifiers

CAS 505-48-6
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00004428
InChI Key TYFQFVWCELRYAO-UHFFFAOYSA-N
Synonym suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h
PubChem CID 10457
ChEBI CHEBI:9300
IUPAC Name octanedioic acid
SMILES OC(=O)CCCCCCC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1396322
View Documents Promotion Details
N/A 100 g
Each for $105.49
Only null left
 
AAA1396336
View Documents Promotion Details
N/A 500 g
Each for $273.33
Only null left
 
AAA139630E
View Documents Promotion Details
N/A 2500 g
Each for $1,009.75
Only null left
 
Description

Description

Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.

Solubility
Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents, reducing agents and bases.
Specifications

Chemical Identifiers

505-48-6
174.20
TYFQFVWCELRYAO-UHFFFAOYSA-N
10457
octanedioic acid
C8H14O4
MFCD00004428
suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h
CHEBI:9300
OC(=O)CCCCCCC(O)=O

Specifications

505-48-6
230°C (15 mmHg)
C8H14O4
1210161
suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h
TYFQFVWCELRYAO-UHFFFAOYSA-N
octanedioic acid
10457
174.2
Suberic acid
141°C to 144°C
203°C (397°F)
MFCD00004428
14,8862
Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform.
OC(=O)CCCCCCC(O)=O
174.20
CHEBI:9300
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 208-010-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only