Learn More
Suberic acid, 99%
CAS: 505-48-6 | C8H14O4 | 174.20 g/mol
Supplier: Thermo Scientific Chemicals A139630E
| Quantity | 2500 g |
|---|
Description
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSuberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.
Solubility
Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform.
Notes
Incompatible with strong oxidizing agents, reducing agents and bases.
Chemical Identifiers
| C8H14O4 | |
| MFCD00004428 | |
| suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h | |
| CHEBI:9300 | |
| OC(=O)CCCCCCC(O)=O |
Specifications
| Suberic acid | |
| 141°C to 144°C | |
| 203°C (397°F) | |
| 2500 g | |
| 1210161 | |
| suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h | |
| TYFQFVWCELRYAO-UHFFFAOYSA-N | |
| octanedioic acid | |
| 10457 | |
| 174.2 |
| 505-48-6 | |
| 230°C (15 mmHg) | |
| C8H14O4 | |
| MFCD00004428 | |
| 14,8862 | |
| Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform. | |
| OC(=O)CCCCCCC(O)=O | |
| 174.20 | |
| CHEBI:9300 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 208-010-9
TSCA : Yes
Recommended Storage : Ambient temperatures