missing translation for 'onlineSavingsMsg'
Learn More

Suberic acid, 99%

Catalog No. AAA1396336
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1396336 500 g
AAA1396322 100 g
AAA139630E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1396336 Supplier Thermo Scientific Chemicals Supplier No. A1396336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 505-48-6 | C8H14O4 | 174.20 g/mol

Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.

Solubility
Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents, reducing agents and bases.

Chemical Identifiers

CAS 505-48-6
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00004428
InChI Key TYFQFVWCELRYAO-UHFFFAOYSA-N
Synonym suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h
PubChem CID 10457
ChEBI CHEBI:9300
IUPAC Name octanedioic acid
SMILES OC(=O)CCCCCCC(O)=O

Specifications

Melting Point 141°C to 144°C
Boiling Point 230°C (15 mmHg)
Flash Point 203°C (397°F)
Quantity 500 g
Beilstein 1210161
Merck Index 14,8862
Solubility Information Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform.
Formula Weight 174.2
Percent Purity 99%
Chemical Name or Material Suberic acid
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 208-010-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only