missing translation for 'onlineSavingsMsg'
Learn More

Strychnine 98.0+%, TCI America™

Supplier:  TCI America S024925G

Catalog No. S024925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Strychnine
C21H22N2O2
25 g
strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos
O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61
334.42
CHEBI:28973
≥98.0% (HPLC,T)
57-24-9
MFCD00005941
1692
QMGVPVSNSZLJIA-FVWCLLPLSA-N
(1R,11S,18S,20R,21R,22S)-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-9-one
441071
334.42
Crystalline Powder

Chemical Identifiers

57-24-9
334.42
QMGVPVSNSZLJIA-FVWCLLPLSA-N
441071
(1R,11S,18S,20R,21R,22S)-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-9-one
C21H22N2O2
MFCD00005941
strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos
CHEBI:28973
O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : WL2275000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions