missing translation for 'onlineSavingsMsg'
Learn More

Strychnine Sulfate Pentahydrate 97.0+%, TCI America™

Catalog No. s009425g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S009425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S009425G Supplier TCI America Supplier No. S009425G

Chemical Identifiers

CAS 60491-10-3
Molecular Formula C42H56N4O13S
Molecular Weight (g/mol) 856.985
MDL Number MFCD00068328
InChI Key FDQUTDUZMOOVST-ZKOSLKCXSA-N
PubChem CID 76965789
IUPAC Name (4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;pentahydrate
SMILES C1CN2CC3=CCOC4CC(=O)N5C6C4C3CC2C61C7=CC=CC=C75.C1CN2CC3=CCOC4CC(=O)N5C6C4C3CC2C61C7=CC=CC=C75.O.O.O.O.O.OS(=O)(=O)O

Specifications

Color White-Yellow
pH 5.5 (1:100 H2O soln.)
Quantity 25 g
UN Number 1692
Formula Weight 766.91
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Strychnine Sulfate Pentahydrate
RTECSNumber WL2550000
TSCA No