missing translation for 'onlineSavingsMsg'
Learn More

Strychnine Hydrochloride Hydrate 98.0+%, TCI America™

Catalog No. S025725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S025725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S025725G Supplier TCI America Supplier No. S025725G
Only null left

Chemical Identifiers

CAS 1421-86-9
Molecular Formula C21H23ClN2O2
Molecular Weight (g/mol) 370.877
MDL Number MFCD00058174
InChI Key VLXYTKMPCOQKEM-ZEYGOCRCSA-N
Synonym strychnine hydrochloride, strychnine hcl, strychnine, monohydrochloride, strychnidin-10-one, monohydrochloride, pubchem8018, c21h22n2o2.hcl, strychnidin-10-one hydrochloride, 10-oxostrychnidin-19-ium chloride, strychnidin-10-one, monohydrochloride 9ci, 4ar,4a1r,5as,8ar,8a1s,15as-4a1,5,5a,7,8,8a1,15,15a-octahydro-2h-4,6-methanoindolo 3,2,1-ij oxepino 2,3,4-de pyrrolo 2,3-h quinolin-14 4ah-one hydrochloride
PubChem CID 16219987
IUPAC Name (4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;hydrochloride
SMILES C1CN2CC3=CCOC4CC(=O)N5C6C4C3CC2C61C7=CC=CC=C75.Cl

Specifications

Melting Point 295°C
Quantity 25 g
UN Number 1692
Formula Weight 370.88
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Strychnine Hydrochloride Hydrate
RTECSNumber WL2430000
TSCA No