accessibility menu, dialog, popup

UserName

NOONCH2HCl

CAS RN 1421-86-9

IUPAC Name: 12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-9-one hydrochloride
Synonyms: strychnos hydrochloride
Molecular Weight (g/mol): 370.88
Molecular Formula: C21H23ClN2O2
InChi Key: VLXYTKMPCOQKEM-UHFFFAOYNA-N
SMILES: Cl.O=C1CC2OCC=C3CN4CCC56C4CC3C2C5N1C1=CC=CC=C61
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results