missing translation for 'onlineSavingsMsg'
Learn More

Strontium AA Standard, 1000 ppm Sr in 3% HCl, Ricca Chemical

$209.53 - $209.53

Chemical Identifiers

CAS 1633-05-2
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
PubChem CID 15407
IUPAC Name strontium(2+) carbonate
SMILES [Sr++].[O-]C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
ASR1KH-500
View Documents
Ricca Chemical Company
ASR1KH500
500 mL Natural Poly Bottle
Each for $209.53
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1633-05-2
147.63
LEDMRZGFZIAGGB-UHFFFAOYSA-L
strontium(2+) carbonate
CO3Sr
MFCD00011250
15407
[Sr++].[O-]C([O-])=O

Specifications

Natural Poly Bottle
94.0,3.45,0.16
98.0,3.59,0.17
Liquid
MFCD00011250
LEDMRZGFZIAGGB-UHFFFAOYSA-L
strontium(2+) carbonate
500 mL
Strontium AA Standard
1633-05-2,7647-01-0,7732-18-5
1,000ppm Sr
1 mL = 1 mg Sr
CO3Sr
Colorless
[Sr++].[O-]C([O-])=O
147.63
15407
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'