missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptomycin Sulfate 95.0+%, TCI America™
$39.87 - $62.66
Chemical Identifiers
| CAS | 3810-74-0 |
|---|---|
| Molecular Formula | C42H84N14O36S3 |
| Molecular Weight (g/mol) | 1457.376 |
| MDL Number | MFCD00037023 |
| InChI Key | QTENRWWVYAAPBI-FFCQDDOVSA-N |
| PubChem CID | 124080941 |
| IUPAC Name | 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-( |
| SMILES | CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O |
Chemical Identifiers
| 3810-74-0 | |
| 1457.376 | |
| QTENRWWVYAAPBI-FFCQDDOVSA-N | |
| 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-( |
| C42H84N14O36S3 | |
| MFCD00037023 | |
| 124080941 | |
| CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O |
Specifications
| Streptomycin Sulfate | |
| 3810-74-0 | |
| C42H84N14O36S3 | |
| QTENRWWVYAAPBI-FFCQDDOVSA-N | |
| 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-( | |
| 124080941 | |
| ≥95.0% (N) |
| Crystalline Powder | |
| White/Yellow | |
| MFCD00037023 | |
| CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O | |
| 1457.376 | |
| 728.69 | |
| Streptomycin Sulfate [for Protein Research] |
Safety and Handling
RTECSNumber : WK4990000
TSCA : Yes