missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptomycin Sulfate 95.0+%, TCI America™
Specifications
Specifications
| Type | Streptomycin Sulfate |
| Physical Form | Crystalline Powder |
| Color | White/Yellow |
| Quantity | 5 g |
| Formula Weight | 728.69 |
| Percent Purity | ≥95.0% (N) |
| Grade | Not Available |
| Chemical Name or Material | Streptomycin Sulfate [for Protein Research] |
Safety and Handling
| RTECSNumber | WK4990000 |
| TSCA | Yes |