missing translation for 'onlineSavingsMsg'
Learn More

Streptomycin Sulfate 95.0+%, TCI America™

Supplier:  TCI America S083425G

Quantity 25 g

 View more versions of this product

Catalog No. S083425G


Only null left

Description

Description

Specifications

Chemical Identifiers

3810-74-0
1457.376
QTENRWWVYAAPBI-FFCQDDOVSA-N
2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
C42H84N14O36S3
MFCD00037023
124080941
CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Streptomycin Sulfate
Crystalline Powder
White/Yellow
C42H84N14O36S3
QTENRWWVYAAPBI-FFCQDDOVSA-N
2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
124080941
≥95.0% (N)
Streptomycin Sulfate [for Protein Research]
3810-74-0
25 g
MFCD00037023
CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O
1457.376
728.69
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : WK4990000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions