missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin, (R-PE)
Streptavidin, (R-PE)
$1234.00 - $1234.00
Specifications
| Concentration | 1 mg/mL |
|---|---|
| Conjugate | R-phycoerythrin |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis |
| Content And Storage | Contains 1 vial of streptavidin,R-PE conjugate. Store in refrigerator (2–8°C). |
Description
- Class: RUO
- Statement: For research use only. Not for use in diagnostic procedures.
- Buffer: PBS
- Preservative: 0.1% Sodium Azide
Specifications
| 1 mg/mL | |
| Fluorescence | |
| Contains 1 vial of streptavidin,R-PE conjugate. Store in refrigerator (2–8°C). |
|
| 488, 488 or 561, 561 | |
| Solution | |
| Streptavidin Conjugate (Fluorescent) |
| R-phycoerythrin | |
| Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis | |
| 488/575 | |
| Wet Ice | |
| PBS Buffer | |
| R-PE |
For Research Use Only. Not for use in diagnostic procedures.