missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (R-PE)

Catalog No. 501121548
Encompass
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121548 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121548 Supplier Invitrogen™ Supplier No. SA10044

Streptavidin, (R-PE)

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide

Specifications

Concentration 1 mg/mL
Conjugate R-phycoerythrin
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity 2 mL
Content And Storage Contains 1 vial of streptavidin,R-PE conjugate.

Store in refrigerator (2–8°C).
Excitation/Emission 488/575
Flow Cytometer Laser Lines 488, 488 or 561, 561
Shipping Condition Wet Ice
Form Solution
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye R-PE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.