missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin Acrylamide

Catalog No. S21379
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S21379 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S21379 Supplier Invitrogen™ Supplier No. S21379

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin acrylamide can be co-polymerized with acrylamide on polymeric surfaces for a uniform monolayer of immobilized streptavidin. Streptavidin acrylamide can also coat beads or microspheres of 1- to 1000-μm diameter to be utilized in a variety of affinity binding or purification applications.

Streptavidin acrylamide can be co-polymerized with acrylamide on polymeric surfaces to create a uniform monolayer of immobilized streptavidin. The streptavidin can then be used for biotinylated-probe isolation, utilizing probes, enzymes, antibodies, or pharmaceutical agents. Streptavidin acrylamide can coat polymeric surfaces such as beads or microspheres that range from 1 to 1000 μm in diameter. The flexibility of using streptavidin acrylamide to coat these surfaces allows for complete customization.

Streptavidin-coated microspheres (microparticles) can be used for a variety of affinity binding applications. Streptavidin rapidly binds to biotin with a strong non-covalent bond and high affinity, allowing this amplified bound complex to remain stable within a range of temperature and pH conditions. This makes streptavidin acrylamide coated microspheres a suitable material for pharmaceutical drug delivery, 3D cell culture, bacterial isolation, and flow cytometry analysis. Streptavidin-coated beads are often utilized for DNA sequencing, PCR product cleanup, cell separation, DNA single molecule imaging studies, and in a variety of diagnostic applications.

Specifications

Content And Storage Store in freezer -5°C to -30°C.
Shipping Condition Room Temperature
Form Liquid Suspension
Quantity 1 mg
For Use With (Application) Cell culture, bacterial isolation, flow cytometry, DNA sequencing, PCR product cleanup, cell separation, DNA single molecule imaging
Type Streptavidin Acrylamide
Label or Dye Acrylamide
Stationary Phase Streptavidin
Column Type Affinity

For Research Use Only. Not for use in diagnostic procedures.