missing translation for 'onlineSavingsMsg'
Learn More

STO-609, 98%, Thermo Scientific Chemicals

Catalog No. AAJ67344LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67344LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67344LB0 Supplier Thermo Scientific Chemicals Supplier No. J67344LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52029-86-4
Molecular Formula C19H10N2O3
Molecular Weight (g/mol) 314.3
MDL Number MFCD06411456
InChI Key MYKOWOGZBMOVBJ-UHFFFAOYSA-N
Synonym 11-oxo-3,10-diazapentacyclo 10.7.1.0 2 , 1 ?.0?,?.0 1 ?, 2 ? icosa-1 20 ,2,4,6,8,12,14,16,18-nonaene-17-carboxylate
PubChem CID 3467590
SMILES C1=CC=C2C(=C1)N=C3N2C(=O)C4=CC=CC5=C(C=CC3=C54)C(=O)O

Specifications

Color Yellow
Quantity 5 mg
Formula Weight 374.35
Percent Purity 98%
Physical Form Solid
Chemical Name or Material STO-609
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only