accessibility menu, dialog, popup

UserName

NNOOHO

CAS RN 52029-86-4

IUPAC Name: 11-oxo-3,10-diazapentacyclo[10.7.1.0²,¹⁰.0⁴,⁹.0¹⁶,²⁰]icosa-1(20),2,4,6,8,12,14,16,18-nonaene-17-carboxylic acid
Synonyms: 11-oxo-3,10-diazapentacyclo[10.7.1.0²,¹⁰.0⁴,⁹.0¹⁶,²⁰]icosa-1(20),2,4,6,8,12,14,16,18-nonaene-17-carboxylic acid
Molecular Weight (g/mol): 314.3
Molecular Formula: C19H10N2O3
InChi Key: MYKOWOGZBMOVBJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=C3C(=O)N4C5=CC=CC=C5N=C4C(C=C1)=C23

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results