missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ Neural Supplement DFS Item

StemPro™ Neural Supplement

Supplier:  Gibco™ A1050801

Catalog No. A1050801


Only null left

Description

Description

Optimal proliferation of scarcily available neural stem and progenitor cells; proliferated cells retain their progenitor status and the potential to diferentiate into neurons and glias. Optimized and ready-to-use system removes the needs for media screening through “trial and error.”

Due to their scarce availability, the in vitro expansion of human neural stem cells (NSC) and glial progenitor cells (GPC) has been a challenge to academic and clinical researchers. To address this challenge, we developed StemPro™ Neural Supplement - an optimal, convenient, and cost-effective serum-free medium supplement for the proliferation of mammalian neural stem and progenitor cells cells, such as human and rat NSCs and GPCs. It can also be used, in combination with appropriate basal medium and growth factors, for the expansion of some neuronal progenitor cells. More importantly, proliferated cells retain their progenitor status and the potential to differentiate into neurons and glial cells.

In recent years, there has been an increased scientific interest in studying Neural Stem⁄Progenitor Cells due to their important roles in neurogenesis. Recent advances have also pointed to the potential use of Neural Stem⁄Progenitor Cells as effective therapies for neurodegenerative diseases, such as dopaminergic neural progenitor cells for Parkinson’s Disease, oligoprogenitor cells for amyotrophic lateral sclerosis (ALS) and multiple sclerosis (MS).
Specifications

Specifications

Store in freezer (-5 to -20°C) and protect from light.

Serum-free
Mammalian Cell Culture
Neural Supplement
Sterile
Academic Research, Clinical Research
10 mL
Serum-Free
Neural Stem Cells
50 X
Liquid
Serum-Free
≤ 50 EU/ml
StemPro
Dry Ice
Endotoxin, Performance, Sterility, pH
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.