missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Staurosporine
Description
TB1285-RMU is an Ancillary Material Grade version of product Staurosporine (Cat. No. 1285). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.
Find out more about our Ancillary Material Grade product range.
Specifications
Specifications
| Chemical Name or Material | [9 S-(9α,10β,11β,13α)]-2,3,10,11,12,13-Hexahydro-10-methoxy-9-methyl-11-(methylamino)-9,13-epoxy-1 H,9 H-diindolo[1,2,3-gh:3 ',2 ',1 '-lm]pyrrolo[3,4-j][1,7 ]benzodiazonin-1-one |
| CAS | 62996-74-1 |
| Quantity | 2 mg |
| Target | Broad Spectrum Protein Kinase Inhibitors |
| Molecular Formula | C28H26N4O3 |
| Purity | 98% |