Learn More
Invitrogen™ STAT6 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA532603
 View more versions of this product
Catalog No. PIPA532603
Only null left
Description
Description
This antibody is predicted to react with canine, mouse, porcine, rabbit based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is placenta or tonsil tissue.
STAT6 protein is a transcription factor activated by cytokines, particularly interleukin-4 and IL13. (Stat6-/-) were found to be deficient in IL-4-mediated functions including Th2 helper T-cell differentiation, expression of cell surface markers, T-cell proliferation, immunoglobulin class switching to IgE, and partial loss of IL-4 mediated proliferation. STAT6 mRNA has been detected in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response. STAT6 is crucial for IL-4-mediated biological responses, and naturally in two isoforms: STAT6a and STAT6c.
Specifications
Chemical Identifiers
STAT6
-20°C, Avoid Freeze/Thaw Cycles
Polyclonal
Liquid
IgG
Human
STAT6
12S1644; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT
STAT6
Primary
Antigen Affinity Chromatography
0.28 mg/mL
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P42226
6778
Synthetic peptide derived from C-terminus of human STAT6.
Antibody
Specifications
STAT6
Polyclonal
Unconjugated
STAT6
12S1644; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT
Rabbit
Antigen Affinity Chromatography
RUO
6778
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot
0.28 mg/mL
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P42226
STAT6
Synthetic peptide derived from C-terminus of human STAT6.
500 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions