Learn More
Thermo Scientific™ StartingBlock™ Blocking Buffer
StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays.
Supplier:  Thermo Scientific™ 37538
 View more versions of this product
Catalog No. PI37538
Only null left
Description
Description
StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays. It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock Blocking Buffer is available in PBS and TBS, with and without Tween 20.
Features of StartingBlock Blocking Buffer include:
  • Strip and re-probe without re-blocking—blots stay blocked even after stripping with Restore Stripping Buffer (Cat. No. 21059)
  • Compatible with many detection systems—western blot, ELISA, and IHC with antibody or avidin/biotin probes (blocker is serum- and biotin-free)
  • Short blocking times—less than 15 minutes for nitrocellulose or PVDF membranes; almost instantaneous for polystyrene microplate wells
Specifications
Chemical Identifiers
Western Blot
Liquid
StartingBlock
Specifications
PBS
Western Blot
StartingBlock
Proprietary protein formulation in phosphate-buffered saline, pH 7.5
1X
Liquid
1 L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Upon receipt store at 4°C.