missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ StartingBlock™ Blocking Buffer

StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays.

Supplier:  Thermo Scientific™ 37539

 View more versions of this product

Catalog No. EN37539


Only null left

Description

Description

StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays. It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock Blocking Buffer is available in PBS and TBS, with and without Tween 20.

Features of StartingBlock Blocking Buffer include:

  • Strip and re-probe without re-blocking—blots stay blocked even after stripping with Restore Stripping Buffer (Cat. No. 21059)
  • Compatible with many detection systems—western blot, ELISA, and IHC with antibody or avidin/biotin probes (blocker is serum- and biotin-free)
  • Short blocking times—less than 15 minutes for nitrocellulose or PVDF membranes; almost instantaneous for polystyrene microplate wells
Specifications

Chemical Identifiers

Western Blot
Liquid
StartingBlock

Specifications

PBST
Western Blot
StartingBlock
Proprietary protein formulation in phosphate-buffered saline, pH 7.5, with 0.05% Tween 20
1X
Liquid
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Upon receipt store at 4°C.

SDS
Documents

Documents

Promotions

Promotions