missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Starter Water Analysis Portable pH Meters

LCD meters combine portability with precise pH measurement.

$1,455.32 - $1,852.64

Specifications

Accuracy (mV) ±1
Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Data Management Details 30 Sets, most recent calibration data
Measurement Modes pH with mV and Temperature
View More Specs

Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
01922081
View Documents Promotion Details
OHAUS™
83033961
ST300
Each for $1,852.64
Only null left
 
01922082
View Documents Promotion Details
OHAUS™
83033962
ST300-B
Each for $1,455.32
Only null left
 
Description

Description

  • Auto buffer recognition stores calibration data to avoid errors
  • Automatic/manual temperature compensation for accurate readings
  • ORP or temperature electrode connectivity for additional functionality
  • IP54 protection shields water/dust damage
  • Size: 90 x 150 x 35 mm
  • Weight: 0.18 kg
Specifications

Specifications

±1
±0.5°C
pH with mV and Temperature
0 to 14
-1999 to 1999
1 to 3
0.1°C
None
Automatic Data Logging
IP54
ABS
AAA
>500 hr.
9 cm
3.5 cm
±0.01
30 Sets, most recent calibration data
Automatic/Manual
0.01
1 mV
0°C to 100°C
LCD
30
BNC, NTC
0.18 kg
0.01 pH; 1 mV; 0.1°C
Portable pH Meters
3.5 in.
1.3 in.
17 cm
Videos
SDS
Documents

Documents