missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Starter Water Analysis Portable pH Meters

Catalog No. 01922081
Change view
Click to view available options
Model:
ST300
ST300-B
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01922081 ST300
01922082 ST300-B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01922081 Supplier OHAUS™ Supplier No. 83033961

LCD meters combine portability with precise pH measurement.

  • Auto buffer recognition stores calibration data to avoid errors
  • Automatic/manual temperature compensation for accurate readings
  • ORP or temperature electrode connectivity for additional functionality
  • IP54 protection shields water/dust damage
  • Size: 90 x 150 x 35 mm
  • Weight: 0.18 kg

Specifications

Data Management Details 30 Sets, most recent calibration data
Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1
Includes 1 Plastic gel electrode, buffer powder sachet
Measurement Modes pH with mV and Temperature
Calibration Automatic/Manual
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) -1999 to 1999
Resolution (mV) 1 mV
Calibration Points 1 to 3
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Display Type LCD
Backlight Option None
Data Points 30
Log Function Type Automatic Data Logging
Inputs BNC, NTC
IP Rating IP54
Weight (Metric) 0.18 kg
Material ABS
Model ST300
Resolution 0.01 pH; 1 mV; 0.1°C
Battery Type AAA
Type Portable pH Meters
Battery Life >500 hr.
Width (English) 3.5 in.
Width (Metric) 9 cm
Height (English) 1.3 in.
Height (Metric) 3.5 cm
Depth (Metric) 17 cm
Show More Show Less