missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Indicator, 0.5 Percent (w/v) Aqueous Solution, Stabilized, Spectrum™ Chemical
Description
Spectrum™ Chemical Starch Indicator, 0.5 Percent (w/v) Aqueous Solution is a ready-for-use solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Specifications
Specifications
| CAS Max % | 0.005, 0.99495, 0.00005 |
| Grade | Chemical Solution |
| Packaging | Poly Bottle |
| Quantity | 500 mL |
| Chemical Name or Material | Starch Indicator |