missing translation for 'onlineSavingsMsg'
Learn More

Starch Indicator, 1% (w/v), with Chloroform preservative, Ricca Chemical

$119.67 - $260.83

Specifications

Chemical Name or Material Starch Indicator
Name Note With Chloroform Preservative
CAS 7732-18-5,9005-84-9,67-66-3
CAS Min % 97.0,0.97,0.1
CAS Max % 100.0,1.01,0.4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
805216
View Documents
Ricca Chemical Company
805216
500 mL Natural Poly Bottle
Each for $119.67
Only null left
 
805232
View Documents
Ricca Chemical Company
805232
1 L Natural Poly Bottle
Each for $156.53
Only null left
 
80521
View Documents
Ricca Chemical Company
80521
4 L Natural Poly Bottle
Each for $260.83
Only null left
 
Description

Description

Specifications

Specifications

Starch Indicator
7732-18-5,9005-84-9,67-66-3
100.0,1.01,0.4
1% W/V
Aqueous Solution
5 to 7
With Chloroform Preservative
97.0,0.97,0.1
1% (w/v)
Indicator
Colorless
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'