missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Indicator, 1% (w/v), with Chloroform preservative, Ricca Chemical
Supplier: Ricca Chemical Company 80521
Specifications
| Starch Indicator | |
| 7732-18-5,9005-84-9,67-66-3 | |
| 100.0,1.01,0.4 | |
| 1% W/V | |
| Natural Poly Bottle | |
| Colorless | |
| 4 L |
| With Chloroform Preservative | |
| 97.0,0.97,0.1 | |
| 1% (w/v) | |
| Indicator | |
| Aqueous Solution | |
| 5 to 7 | |
| Liquid |