missing translation for 'onlineSavingsMsg'
Learn More

Starch Indicator, 1% (w/v), with Chloroform preservative, Ricca Chemical

Supplier:  Ricca Chemical Company 805216

 View more versions of this product

Catalog No. 805216


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Starch Indicator
7732-18-5 , 9005-84-9 , 67-66-3
100.0,1.01,0.4
1% W/V
Natural Poly Bottle
Colorless
500 mL
With Chloroform Preservative
97.0,0.97,0.1
1% (w/v)
Indicator
Aqueous Solution
5 to 7
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions