missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride Reagent I, For Phosphorus, Certified, LabChem™

Supplier:  LabChem LC252301

Catalog No. LC252301

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Stannous Chloride Reagent I
For Phosphorus
97.5,2.5
SnCl2·2H2O
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
225.65
Passes Test
Passes Test
500 mL
Colorless
56-81-5 , 10025-69-1
C3H8O3
MFCD00004722
Soluble in water
OCC(O)CO
92.09
CHEBI:17754
Certified
Poly Bottle
Hydrogen chloride; Carbon monoxide; Carbon dioxide
Liquid

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation and serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions