missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, 40% (w/v) in concentrated Hydrochloric Acid, Ricca Chemical

Catalog No. p-3252450
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
79954 120 mL Amber Glass Bottle
799516 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 79954 Supplier Ricca Chemical Company Supplier No. 79954
Only null left

Chemical Identifiers

CAS 7647-01-0 , 10025-69-1
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless to Green-Yellow
CAS Min % 73.0, 24.82
CAS Max % 76.0, 25.84
Percent Purity 40%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Concentrated Hydrochloric Acid
pH <1
Quantity 120 mL
Concentration or Composition Notes 40% (w/v)
Chemical Name or Material Stannous Chloride
Show More Show Less