missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, 10% (w/v) in 20% (v/v) Hydrochloric Acid, Ricca Chemical

Catalog No. 7996416
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7996416 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7996416 Supplier Ricca Chemical Company Supplier No. 7996.416

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 10025-69-1 , 7647-01-0
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Colorless
CAS Min % 73.0, 17.31, 7.34
CAS Max % 76.0, 18.02, 7.64
Physical Form Liquid
Quantity 500 mL
Percent Purity 10% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) in 20% (v/v) Hydrochloric Acid
Packaging Amber Glass Bottle
Chemical Name or Material Stannous Chloride
ShelfLife 60 days