Learn More
Thermo Scientific™ Stands for 1300 Series A2 Biological Safety Cabinets
Electric and manual adjustable and rolling castor stands
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Product Type Height (English) Exterior Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Product Type Height (English) Exterior Price Quantity & Availability  
13261350
View Documents Promotion Details
Thermo Scientific™
1911410
5 ft. Cabinet Rolling Caster Stand 30 in. N/A
Only null left
13261349
View Documents Promotion Details
Thermo Scientific™
1911409
3 ft. Cabinet Rolling Caster Stand 30 in. N/A
Only null left
13261348
View Documents Promotion Details
Thermo Scientific™
1911401
5 ft. Cabinet Adjustable Stand 30 to 38 in. N/A
Only null left
13261347
View Documents Promotion Details
Thermo Scientific™
1911400
3 ft. Cabinet Adjustable Stand 30 to 38 in. N/A
Only null left
Description
Description
Electric and manual adjustable and rolling castor stands
  • Manual and electric stands provide comfortable work surface height of 30 to 38 in. for seated or standing applications
  • Manual stands are adjustable in 2 in. increments; electric stands are infinitely adjustable
  • Rolling caster stand provides comfortable fixed worksurface height of 30 in. for seated applications; designed for limited cabinet mobility for cleaning purposes
  • Stands are included with packages